accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(R)OH3CO(S)OH(R)OH(S)CH3O(R)OHH3CH3C

CAS RN 60857-08-1

IUPAC Name: (1R,2S,6R,10S,15R)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-13-yl acetate
Synonyms: prostratin
Molecular Weight (g/mol): 390.48
Molecular Formula: C22H30O6
InChi Key: BOJKFRKNLSCGHY-KKFGZBDXSA-N
SMILES: C[C@@H]1CC2(OC(C)=O)C([C@@H]3C=C(CO)C[C@@]4(O)[C@@H](C=C(C)C4=O)[C@@]13O)C2(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results