accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6086-42-6

IUPAC Name: 4,4-dimethylimidazolidine-2-thione
Synonyms: 4,4-dimethylimidazolidine-2-thione
Molecular Weight (g/mol): 130.21
Molecular Formula: C5H10N2S
InChi Key: TXRCQKVSGVJWAW-UHFFFAOYSA-N
SMILES: CC1(C)CNC(=S)N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results