Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4-Dimethyl-2-imidazolidinethione 98.0+%, TCI America™
SDP

Catalog No. D243510G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D243510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D243510G Supplier TCI America Supplier No. D243510G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6086-42-6
Molecular Formula C5H10N2S
Molecular Weight (g/mol) 130.209
MDL Number MFCD00661072
InChI Key TXRCQKVSGVJWAW-UHFFFAOYSA-N
Synonym 4,4-Dimethylethylenethiourea, 2-Mercapto-4,4-dimethylimidazoline
PubChem CID 3032502
IUPAC Name 4,4-dimethylimidazolidine-2-thione
SMILES CC1(CNC(=S)N1)C

Specifications

Melting Point 120°C
Color White
Quantity 10 g
Formula Weight 130.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4-Dimethyl-2-imidazolidinethione
RTECSNumber NI9690000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.