accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6087-56-5

IUPAC Name: boron(3+) 4-(dimethylamino)benzene-1-diazonium tetrafluoride
Synonyms: boron(3+) 4-(dimethylamino)benzenediazonium tetrafluoride
Molecular Weight (g/mol): 234.99
Molecular Formula: C8H10BF4N3
InChi Key: FNXHWKBYOXDBCH-UHFFFAOYSA-J
SMILES: [B+3].[F-].[F-].[F-].[F-].CN(C)C1=CC=C(C=C1)[N+]#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results