Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Diazo-N,N-dimethylaniline Chloride Zinc Chloride Hydrate 95.0+%, TCI America™
SDP

Catalog No. D0140500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0140500G 500 g
D014025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0140500G Supplier TCI America Supplier No. D0140500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6087-56-5 , 13533-17-0
Molecular Formula C8H10Cl3N3Zn
Molecular Weight (g/mol) 319.919
MDL Number MFCD00031685
InChI Key SIMFPAVQCFHVGM-UHFFFAOYSA-K
Synonym N,N-Dimethyl-1,4-phenylenediamine Diazonium Chloride Zinc Chloride
PubChem CID 73555201
IUPAC Name 4-(dimethylamino)benzenediazonium;trichlorozinc(1-)
SMILES CN(C)C1=CC=C(C=C1)[N+]#N.Cl[Zn-](Cl)Cl

Specifications

Quantity 500 g
UN Number 1325
Formula Weight 319.92
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Diazo-N,N-dimethylaniline Chloride Zinc Chloride Hydrate
EINECSNumber (3)-0253
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.