accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBrCH3Br

CAS RN 609-11-0

IUPAC Name: ethyl 2,3-dibromobutanoate
Synonyms: ethyl 2,3-dibromobutanoate
Molecular Weight (g/mol): 273.95
Molecular Formula: C6H10Br2O2
InChi Key: YPZLAXFGOUPECS-UHFFFAOYNA-N
SMILES: CCOC(=O)C(Br)C(C)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results