Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2,3-Dibromobutyrate 95.0+%, TCI America™
SDP

Catalog No. D26125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26125G Supplier TCI America Supplier No. D26125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 609-11-0
Molecular Formula C6H10Br2O2
Molecular Weight (g/mol) 273.952
MDL Number MFCD00017863
InChI Key YPZLAXFGOUPECS-UHFFFAOYSA-N
Synonym 2,3-Dibromobutyric Acid Ethyl Ester
PubChem CID 98157
IUPAC Name ethyl 2,3-dibromobutanoate
SMILES CCOC(=O)C(C(C)Br)Br

Specifications

Color Yellow
Boiling Point 104°C
Quantity 5 g
UN Number 1760
Formula Weight 273.95
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl 2,3-Dibromobutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.