accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6092-54-2

IUPAC Name: hexyl carbonochloridate
Synonyms: hexyl carbonochloridate
Molecular Weight (g/mol): 164.63
Molecular Formula: C7H13ClO2
InChi Key: KIWBRXCOTCXSSZ-UHFFFAOYSA-N
SMILES: CCCCCCOC(Cl)=O
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)