Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexyl chloroformate

Catalog No. 2527785G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-277-85G 5 g
25-277-825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-277-85G Supplier Sigma Aldrich Supplier No. 2527785G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Hexyl chloroformate has been used as derivatization reagent in:
  • Determination of sarcosine and n-ethylglycine in urine and urinary sediments by solid-phase micro extraction and fast GC-ms analysis
  • Determination of benzoylecgonine in urine by GC-quadrupole ion trap mass spectrometry

Specifications

CAS 6092-54-2
Density 1.007 g/mL (at 25°C (literature))
Boiling Point 60°C to 61°C (7 mmHg)
Molecular Formula C7H13ClO2
Refractive Index n20/D 1.424 (literature)
Linear Formula ClCO2(CH2)5 CH3
MDL Number MFCD00000651
Quantity 5 g
Molecular Weight (g/mol) 164.63
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.