accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OFBOHHO

CAS RN 609807-25-2

IUPAC Name: (3-fluoro-5-methoxyphenyl)boronic acid
Synonyms: 3-fluoro-5-methoxyphenylboronic acid
Molecular Weight (g/mol): 169.95
Molecular Formula: C7H8BFO3
InChi Key: MQFKLCWETPKBCH-UHFFFAOYSA-N
SMILES: COC1=CC(F)=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results