accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNHOHHO

CAS RN 61-76-7

IUPAC Name: hydrogen 3-[1-hydroxy-2-(methylamino)ethyl]phenol chloride
Synonyms: DL-phenylephrine hydrochloride
Molecular Weight (g/mol): 203.67
Molecular Formula: C9H14ClNO2
InChi Key: OCYSGIYOVXAGKQ-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CNCC(O)C1=CC=CC(O)=C1
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (2)
  • (4)
  • (3)
Grade 
  • (1)
  • (4)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (5)
  • (2)
  • (4)
  • (3)
  • (1)
  • (4)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)