accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO(S)(R)OO

CAS RN 610-09-3

IUPAC Name: (1R,2S)-cyclohexane-1,2-dicarboxylate
Synonyms: (1R,2S)-cyclohexane-1,2-dicarboxylate
Molecular Weight (g/mol): 170.17
Molecular Formula: C8H10O4
InChi Key: QSAWQNUELGIYBC-OLQVQODUSA-L
SMILES: [O-]C(=O)[C@H]1CCCC[C@H]1C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)