Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,2-Cyclohexanedicarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. C0458500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0458500G 500 g
C045825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0458500G Supplier TCI America Supplier No. C0458500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 610-09-3
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.17
MDL Number MFCD00064949
InChI Key QSAWQNUELGIYBC-OLQVQODUSA-L
Synonym cis-1,2-cyclohexanedicarboxylic acid, cis-cyclohexane-1,2-dicarboxylic acid, cis-hexahydrophthalic acid, 1r,2s-cyclohexane-1,2-dicarboxylic acid, 1s,2r-cyclohexane-1,2-dicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, 1r,2s-rel, cis-1,2-cyclohexanedicarboxylicacid, ksc497o9d, cis-+/--perhydrophthalic acid, cis-cyclohexane-1,2-dicarboxylicacid
PubChem CID 245564
IUPAC Name (1R,2S)-cyclohexane-1,2-dicarboxylate
SMILES [O-]C(=O)[C@H]1CCCC[C@H]1C([O-])=O

Specifications

Melting Point 198°C
Color White
Quantity 500 g
Formula Weight 172.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material cis-1,2-Cyclohexanedicarboxylic Acid
EINECSNumber (3)-2678
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.