CAS RN 610-27-5
4-Nitrophthalic acid, 97%
CAS: 610-27-5 Molecular Formula: C8H5NO6 Molecular Weight (g/mol): 211.129 MDL Number: MFCD00007252 InChI Key: SLBQXWXKPNIVSQ-UHFFFAOYSA-N Synonym: 4-nitrophthalic acid,5-nitrophthalic acid,4-nitrobenzene-1,2-dicarboxylic acid,1,2-benzenedicarboxylic acid, 4-nitro,phthalic acid, 4-nitro,4-nitrophthalicacid,p-nitrophthalic acid,4-nitro-phthalic acid,acmc-209mo8 PubChem CID: 69121 SMILES: C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)C(=O)O
4-Nitrophthalic acid, 99%
CAS: 610-27-5 Molecular Formula: C8H5NO6 Molecular Weight (g/mol): 211.13 InChI Key: SLBQXWXKPNIVSQ-UHFFFAOYSA-N Synonym: 4-nitrophthalic acid,5-nitrophthalic acid,4-nitrobenzene-1,2-dicarboxylic acid,1,2-benzenedicarboxylic acid, 4-nitro,phthalic acid, 4-nitro,4-nitrophthalicacid,p-nitrophthalic acid,4-nitro-phthalic acid,acmc-209mo8 PubChem CID: 69121 SMILES: C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)C(=O)O
4-Nitrophthalic acid, MP Biomedicals™
CAS: 610-27-5 Molecular Formula: C8H5NO6 Molecular Weight (g/mol): 211.13 MDL Number: MFCD00007252 InChI Key: SLBQXWXKPNIVSQ-UHFFFAOYSA-N
4-Nitrophthalic Acid 98.0+%, TCI America™
CAS: 610-27-5 Molecular Formula: C8H5NO6 Molecular Weight (g/mol): 211.129 MDL Number: MFCD00007252 InChI Key: SLBQXWXKPNIVSQ-UHFFFAOYSA-N Synonym: 4-nitrophthalic acid,5-nitrophthalic acid,4-nitrobenzene-1,2-dicarboxylic acid,1,2-benzenedicarboxylic acid, 4-nitro,phthalic acid, 4-nitro,4-nitrophthalicacid,p-nitrophthalic acid,4-nitro-phthalic acid,acmc-209mo8 PubChem CID: 69121 SMILES: C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)C(=O)O
Sigma Aldrich 4-Nitrophthalic acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.