Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophthalic acid, MP Biomedicals™

Catalog No. MP215592080 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215592080 100 g
MP215592010 10 g
MP215592025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP215592080 Supplier MP Biomedicals, Inc Supplier No. 0215592080
Add to Cart
Add to Cart

4-Nitrophthalic acid 90%, Pale Yellow, Crystalline Powder

  • It was used in the preparation of a 2D homochiral inorganic-organic framework {[Mn(4-nitrophthalate)(4,4′-bipyridine)(H2O)]·(H2O2}n by reacting with manganese ions, and ancillary 4,4′-bipyridine ligands.
  • The tissue-specific activity of 4-nitrophthalic acid was studied.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-27-5
Molecular Formula C8H5NO6
Molecular Weight (g/mol) 211.13
MDL Number MFCD00007252
InChI Key SLBQXWXKPNIVSQ-UHFFFAOYSA-N

Specifications

Melting Point 159°C to 161°C (literature)
Color Pale Yellow
Loss on Drying 0.0028
Quantity 100 g
Beilstein 1882262
Grade 0.9
storageNote1 Room Temperatur
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.