accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 610-36-6

IUPAC Name: 4-amino-2-nitrobenzoic acid
Synonyms: 4-amino-2-nitrobenzoic acid
Molecular Weight (g/mol): 182.14
Molecular Formula: C7H6N2O4
InChi Key: SAJYSJVBNGUWJK-UHFFFAOYSA-N
SMILES: NC1=CC=C(C(O)=O)C(=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results