Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-nitrobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. A16861G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A16861G Supplier TCI America Supplier No. A16861G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 610-36-6
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.135
InChI Key SAJYSJVBNGUWJK-UHFFFAOYSA-N
PubChem CID 235713
SMILES C1=CC(=C(C=C1N)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 239°C
Quantity 1 g
Formula Weight 182.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-nitrobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.