accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 610-66-2

IUPAC Name: 2-(2-nitrophenyl)acetonitrile
Synonyms: benzeneacetonitrile, 2-nitro-
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: YPRFCQAWSNWRLM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1CC#N
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)