Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Nitrobenzyl Cyanide 98.0+%, TCI America™
SDP

Catalog No. N07815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07815G 5 g
N078125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07815G Supplier TCI America Supplier No. N07815G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 610-66-2
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00007183
InChI Key YPRFCQAWSNWRLM-UHFFFAOYSA-N
Synonym 2-nitrophenylacetonitrile, 2-2-nitrophenyl acetonitrile, 2-nitrophenyl acetonitrile, benzeneacetonitrile, 2-nitro, 2-nitrobenzyl cyanide, o-nitrophenyl acetonitrile, o-nitrobenzacetonitrile, 2-nitrobenzeneacetonitrile, 2-nitrobenzyl nitrile, acetonitrile, o-nitrophenyl
PubChem CID 11888
IUPAC Name 2-(2-nitrophenyl)acetonitrile
SMILES [O-][N+](=O)C1=CC=CC=C1CC#N

Specifications

Melting Point 84°C
Boiling Point 178°C
Quantity 5 g
UN Number 3439
Formula Weight 162.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrobenzyl Cyanide
EINECSNumber (3)-1820
RTECSNumber AM1248000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.