CAS RN 610-67-3
IUPAC Name:
1-ethoxy-2-nitrobenzene
Synonyms:
benzene, 1-ethoxy-2-nitro-
Molecular Weight (g/mol):
167.16
Molecular Formula:
C8H9NO3
InChi Key:
XGLGESCVNJSAQY-UHFFFAOYSA-N
SMILES:
CCOC1=CC=CC=C1[N+]([O-])=O
1
–
1
of
1
results
1
2-Nitrophenetole 98.0+%, TCI America™
CAS: 610-67-3 Molecular Formula: C8H9NO3 Molecular Weight (g/mol): 167.16 MDL Number: MFCD00053603 InChI Key: XGLGESCVNJSAQY-UHFFFAOYSA-N Synonym: 1-Ethoxy-2-nitrobenzene PubChem CID: 11889 IUPAC Name: 1-ethoxy-2-nitrobenzene SMILES: CCOC1=CC=CC=C1[N+]([O-])=O