CAS RN 6100-05-6
Tripotassium citrate monohydrate, 99+%
CAS: 6100-05-6 Molecular Formula: C6H7K3O8 Molecular Weight (g/mol): 324.41 MDL Number: MFCD00150442 InChI Key: PJAHUDTUZRZBKM-UHFFFAOYSA-K Synonym: potassium citrate monohydrate,tripotassium citrate monohydrate,unii-ee90oni6ff,ee90oni6ff,citric acid tripotassium salt,urocit-k,ccris 6543,tri-potassium citrate monohydrate,potassium citrate tribasic monohydrate,citric acid, tripotassium salt, monohydrate PubChem CID: 2735208 ChEBI: CHEBI:64746 IUPAC Name: tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate SMILES: C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
Tripotassium citrate monohydrate, 97%
CAS: 6100-05-6 Molecular Formula: C6H7K3O8 Molecular Weight (g/mol): 324.41 MDL Number: MFCD00150442 InChI Key: PJAHUDTUZRZBKM-UHFFFAOYSA-K Synonym: potassium citrate monohydrate,tripotassium citrate monohydrate,unii-ee90oni6ff,ee90oni6ff,citric acid tripotassium salt,urocit-k,ccris 6543,tri-potassium citrate monohydrate,potassium citrate tribasic monohydrate,citric acid, tripotassium salt, monohydrate PubChem CID: 2735208 ChEBI: CHEBI:64746 IUPAC Name: tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate SMILES: C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
Potassium citrate tribasic monohydrate, 99%, extra pure
CAS: 6100-05-6 Molecular Formula: C6H5K3O7·H2O Molecular Weight (g/mol): 324.42 InChI Key: PJAHUDTUZRZBKM-UHFFFAOYSA-K Synonym: potassium citrate monohydrate,tripotassium citrate monohydrate,unii-ee90oni6ff,ee90oni6ff,citric acid tripotassium salt,urocit-k,ccris 6543,tri-potassium citrate monohydrate,potassium citrate tribasic monohydrate,citric acid, tripotassium salt, monohydrate PubChem CID: 2735208 ChEBI: CHEBI:64746 IUPAC Name: tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate SMILES: C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
Potassium Citrate, Monohydrate, Granular, U.S.P. - F.C.C., J.T. Baker™
CAS: 6100-05-6 Molecular Formula: C6H7K3O8 Molecular Weight (g/mol): 324.41 InChI Key: PJAHUDTUZRZBKM-UHFFFAOYSA-K Synonym: potassium citrate monohydrate,tripotassium citrate monohydrate,unii-ee90oni6ff,ee90oni6ff,citric acid tripotassium salt,urocit-k,ccris 6543,tri-potassium citrate monohydrate,potassium citrate tribasic monohydrate,citric acid, tripotassium salt, monohydrate PubChem CID: 2735208 ChEBI: CHEBI:64746 IUPAC Name: tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate SMILES: C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
Potassium Citrate, Monohydrate, Granular, USP, 99-100.5%, Spectrum™ Chemical
CAS: 6100-05-6 Molecular Formula: C6H7K3O8 Molecular Weight (g/mol): 324.41 InChI Key: PJAHUDTUZRZBKM-UHFFFAOYSA-K IUPAC Name: tripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate SMILES: O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Potassium Citrate, Monohydrate, Crystal, 99%, Spectrum™ Chemical
CAS: 6100-05-6
Potassium Citrate, Monohydrate, Granular, BAKER ANALYZED™ Reagent, J.T. Baker™
CAS: 6100-05-6 Molecular Formula: C6H7K3O8 Molecular Weight (g/mol): 324.41 InChI Key: PJAHUDTUZRZBKM-UHFFFAOYSA-K Synonym: potassium citrate monohydrate,tripotassium citrate monohydrate,unii-ee90oni6ff,ee90oni6ff,citric acid tripotassium salt,urocit-k,ccris 6543,tri-potassium citrate monohydrate,potassium citrate tribasic monohydrate,citric acid, tripotassium salt, monohydrate PubChem CID: 2735208 ChEBI: CHEBI:64746 IUPAC Name: tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate SMILES: C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
Potassium Citrate Monohydrate, Reagent (Tripotassium Citrate Monohydrate), Reagents
CAS: 6100-05-6 Molecular Formula: C6H7K3O8 Molecular Weight (g/mol): 324.41 InChI Key: PJAHUDTUZRZBKM-UHFFFAOYSA-K Synonym: Tripotassium Citrate Monohydrate IUPAC Name: tripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate SMILES: O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Medisca Potassium Citrate, USP (Dietary Supplement Grade)
Potassium Citrate, USP (Dietary Supplement Grade) comes as either transparent crystals or as a white, granular powder. It is deliquescent when exposed to moist air.
Sigma Aldrich Potassium Citrate Tribasic Monohydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
tri-Potassium citrate monohydrate EMPROVE™ ESSENTIAL Ph Eur, BP, USP, FCC, E 332, 100.5%, MilliporeSigma™
CAS: 6100-05-6 Molecular Formula: C6H7K3O8 Molecular Weight (g/mol): 324.41 InChI Key: PJAHUDTUZRZBKM-UHFFFAOYSA-K Synonym: Tripotassium citrate,Citric acid potassium salt,2-Hydroxy-1,2,3-Propanetriccarboxylic Acid IUPAC Name: tripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate SMILES: O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O