Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Water Standard KF-Oven 220°C-230°C, Standard for Karl Fischer oven check (water content ∼5.55%), Solstice

Catalog No. 6002553 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-025-53 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-025-53 Supplier Solstice Supplier No. 347486X10G
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Standard for Karl Fischer oven check (water content ∼5.55%, exact value on report of analysis)

Advantages of HYDRANAL-Water Standards:

  • Broad product range for volumetric and coulometric Karl Fischer applications, as well as the indirect oven application
  • Manufactured according to ISO 9001 requirements
  • Report of Analysis showing the exact water content and expanded uncertainty

Verified against external standards such as NIST SRM 2890 or NMIJ CRM 4222

  • Long shelf life (up to five years)
  • Convenient packaging
  • Supplied with detailed instruction for use
TRUSTED_SUSTAINABILITY

Specifications

CAS 6100-05-6
Quantity 10 g
Packaging Glass bottle
UN Number NONH for all modes of transport
Chemical Name or Material HYDRANAL™-Water Standard KF-Oven 220°C-230°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.