accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClCl

CAS RN 61023-66-3

IUPAC Name: 1-(2,4-dichlorophenyl)pentan-1-one
Synonyms: 1-(2,4-dichlorophenyl)pentan-1-one
Molecular Weight (g/mol): 231.12
Molecular Formula: C11H12Cl2O
InChi Key: XVWXSWROOLWNCJ-UHFFFAOYSA-N
SMILES: CCCCC(=O)C1=C(Cl)C=C(Cl)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results