Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',4'-Dichlorovalerophenone 95.0+%, TCI America™
SDP

Catalog No. D33665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33665G 5 g
D336625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33665G Supplier TCI America Supplier No. D33665G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61023-66-3
Molecular Formula C11H12Cl2O
Molecular Weight (g/mol) 231.116
InChI Key XVWXSWROOLWNCJ-UHFFFAOYSA-N
PubChem CID 10988086
IUPAC Name 1-(2,4-dichlorophenyl)pentan-1-one
SMILES CCCCC(=O)C1=C(C=C(C=C1)Cl)Cl

Specifications

Quantity 5 g
Synonym 2′C,4′C-Dichloropentanophenone
Formula Weight 231.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,4′-Dichlorovalerophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.