accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61034-86-4

IUPAC Name: [2-(1H-pyrrol-1-yl)phenyl]methanol
Synonyms: [2-(pyrrol-1-yl)phenyl]methanol
Molecular Weight (g/mol): 173.22
Molecular Formula: C11H11NO
InChi Key: PMFMGYSILUCETA-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1N1C=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results