Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[2-(1H-Pyrrol-1-yl)phenyl]methanol, 97%, Thermo Scientific™

Catalog No. CC34309EA
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC34309EA 10 g
CC34309CB 250 mg
CC34309DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC34309EA Supplier Thermo Scientific Chemicals Supplier No. CC34309EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61034-86-4
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.215
MDL Number MFCD04115123
InChI Key PMFMGYSILUCETA-UHFFFAOYSA-N
Synonym 2-1h-pyrrol-1-yl phenyl methanol, 2-1h-pyrrol-1-yl benzyl alcohol, 2-pyrrol-1-yl-phenyl-methanol, 2-pyrrol-1-yl phenyl methanol, 2-1h-pyrrol-1-yl benzylalcohol, pubchem11305, 2-pyrrolylphenyl methan-1-ol, 2-pyrrol-1-yl-benzyl alcohol, 1-2-hydroxymethyl phenyl-1h-pyrrole
PubChem CID 7016493
IUPAC Name (2-pyrrol-1-ylphenyl)methanol
SMILES C1=CC=C(C(=C1)CO)N2C=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 173.21
Percent Purity 97%
Chemical Name or Material [2-(1H-Pyrrol-1-yl)phenyl]methanol
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.