accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61040-81-1

IUPAC Name: 3,5-dimethoxy-4-methylbenzoate
Synonyms: 3,5-dimethoxy-4-methylbenzoate
Molecular Weight (g/mol): 195.2
Molecular Formula: C10H11O4
InChi Key: QIBMVRYNEXOCCF-UHFFFAOYSA-M
SMILES: COC1=CC(=CC(OC)=C1C)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results