CAS RN 6105-75-5
CAS RN 6105-75-5
IUPAC Name:
2-methyl-1-(morpholin-4-yl)propan-2-amine
Synonyms:
2-methyl-1-(morpholin-4-yl)propan-2-amine
Molecular Weight (g/mol):
158.25
Molecular Formula:
C8H18N2O
InChi Key:
JCXYZQUYVNLCTG-UHFFFAOYSA-N
SMILES:
CC(C)(N)CN1CCOCC1
1
–
1
of
1
results
1
2-Methyl-1-morpholino-2-propanamine, 97%, Thermo Scientific™
CAS: 6105-75-5 Molecular Formula: C8H18N2O Molecular Weight (g/mol): 158.25 MDL Number: MFCD01714129 InChI Key: JCXYZQUYVNLCTG-UHFFFAOYSA-N Synonym: 2-methyl-1-morpholinopropan-2-amine,2-methyl-1-morpholin-4-yl propan-2-amine,1,1-dimethyl-2-morpholin-4-yl-ethylamine,2-methyl-1-morpholino-2-propanamine,4-2-amino-2-methylpropyl morpholine,morpholine, 4-2-amino-2-methylpropyl,morpholine, n-2-aminoisobutyl,1,1-dimethyl-2-morpholin-4-ylethylamine,1,1-dimethyl-2-morpholin-4-ylethyl amine,4-morpholineethanamine, alpha,alpha-dimethyl PubChem CID: 201497 IUPAC Name: 2-methyl-1-morpholin-4-ylpropan-2-amine SMILES: CC(C)(N)CN1CCOCC1