accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2ONaNH2(S)OOOHO

CAS RN 6106-04-3

IUPAC Name: sodium (4S)-4-amino-4-carboxybutanoate hydrate
Synonyms: sodium (4S)-4-amino-4-carboxybutanoate hydrate
Molecular Weight (g/mol): 187.13
Molecular Formula: C5H10NNaO5
InChi Key: GJBHGUUFMNITCI-QTNFYWBSSA-M
SMILES: O.[Na+].N[C@@H](CCC([O-])=O)C(O)=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (1)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)