Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamic acid monosodium salt monohydrate, 98+%

Catalog No. AAA129190B
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA129190B 1000 g
AAA1291930 250 g
AAA129190I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA129190B Supplier Thermo Scientific Chemicals Supplier No. A129190B
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6106-04-3 | C5H10NNaO5 | 187.13 g/mol

L-Glutamic acid monosodium salt monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. Also used to intensify flavors of foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid monosodium salt monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. Also used to intensify flavors of foods.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-04-3
Molecular Formula C5H10NNaO5
Molecular Weight (g/mol) 187.13
MDL Number MFCD00150138
InChI Key GJBHGUUFMNITCI-QTNFYWBSSA-M
Synonym monosodium glutamate, l-glutamic acid monosodium salt monohydrate, p.a, l-glutamic acid monosodium salt monohydrate nt, l-glutamic acid monosodium salt monohydrate, saj special grade, monosodium glutamate, united states pharmacopeia usp reference standard, l-glutamic acid, monosodium salt, low endotoxin, pharmagrade, manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., suitable for cell culture
PubChem CID 87090819
IUPAC Name (2S)-2-aminopentanedioic acid;sodium;hydrate
SMILES C(CC(=O)O)C(C(=O)O)N.O.[Na]

Specifications

Melting Point 228°C (decomposition)
Beilstein 4164348
Solubility Information Soluble in water.
Quantity 1000 g
Formula Weight 187.14 (169.12 Anhydrous)
Optical Rotation +22° (c=1 in 6N HCl)
Percent Purity ≥98%
Chemical Name or Material L-Glutamic acid monosodium salt monohydrate
EINECSNumber 205-538-1
RTECSNumber MA1578000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.