accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONOOCH3

CAS RN 610791-05-4

IUPAC Name: 1-tert-butyl 3-methyl azetidine-1,3-dicarboxylate
Synonyms: 1-tert-butyl 3-methyl azetidine-1,3-dicarboxylate
Molecular Weight (g/mol): 215.25
Molecular Formula: C10H17NO4
InChi Key: SECFRXGVLMVUPD-UHFFFAOYSA-N
SMILES: COC(=O)C1CN(C1)C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results