CAS RN 6108-17-4
Lithium acetate dihydrate, 98%, extra pure
CAS: 6108-17-4 Molecular Formula: C2H3LiO2·2H2O Molecular Weight (g/mol): 102.02 MDL Number: MFCD00066949 InChI Key: IAQLJCYTGRMXMA-UHFFFAOYSA-M Synonym: lithium acetate dihydrate,lithiumacetatedihydrate,lithium acetate dihydrate, reagent grade,lioac-2h2o,lioac.2h2o,ksc498a1d,c2h3o2.li.2h2o,lithotab acetate ion dihydrate,acetic acid lithium salt dihydrate PubChem CID: 23666338 IUPAC Name: lithium;acetate;dihydrate SMILES: [Li+].CC(=O)[O-].O.O
Lithium acetate dihydrate, MP Biomedicals™
CAS: 6108-17-4 Molecular Formula: C2H7LiO4 Molecular Weight (g/mol): 102.014 InChI Key: IAQLJCYTGRMXMA-UHFFFAOYSA-M Synonym: lithium acetate dihydrate,lithiumacetatedihydrate,lithium acetate dihydrate, reagent grade,lioac-2h2o,lioac.2h2o,ksc498a1d,c2h3o2.li.2h2o,lithotab acetate ion dihydrate,acetic acid lithium salt dihydrate PubChem CID: 23666338 IUPAC Name: lithium;acetate;dihydrate SMILES: [Li+].CC(=O)[O-].O.O
Lithium acetate dihydrate, Reagent Grade
CAS: 6108-17-4 Molecular Formula: C2H7LiO4 Molecular Weight (g/mol): 102.014 MDL Number: MFCD00066949 InChI Key: IAQLJCYTGRMXMA-UHFFFAOYSA-M Synonym: lithium acetate dihydrate,lithiumacetatedihydrate,lithium acetate dihydrate, reagent grade,lioac-2h2o,lioac.2h2o,ksc498a1d,c2h3o2.li.2h2o,lithotab acetate ion dihydrate,acetic acid lithium salt dihydrate PubChem CID: 23666338 IUPAC Name: lithium;acetate;dihydrate SMILES: [Li+].CC(=O)[O-].O.O
Lithium acetate dihydrate, 98%, for analysis
CAS: 6108-17-4 Molecular Formula: C2H3LiO2·2H2O Molecular Weight (g/mol): 102.02 InChI Key: IAQLJCYTGRMXMA-UHFFFAOYSA-M Synonym: lithium acetate dihydrate,lithiumacetatedihydrate,lithium acetate dihydrate, reagent grade,lioac-2h2o,lioac.2h2o,ksc498a1d,c2h3o2.li.2h2o,lithotab acetate ion dihydrate,acetic acid lithium salt dihydrate PubChem CID: 23666338 IUPAC Name: lithium;acetate;dihydrate SMILES: [Li+].CC(=O)[O-].O.O
Lithium acetate dihydrate, 99%
CAS: 6108-17-4 Molecular Formula: C2H7LiO4 Molecular Weight (g/mol): 102.014 MDL Number: MFCD00066949 InChI Key: IAQLJCYTGRMXMA-UHFFFAOYSA-M Synonym: lithium acetate dihydrate,lithiumacetatedihydrate,lithium acetate dihydrate, reagent grade,lioac-2h2o,lioac.2h2o,ksc498a1d,c2h3o2.li.2h2o,lithotab acetate ion dihydrate,acetic acid lithium salt dihydrate PubChem CID: 23666338 IUPAC Name: lithium;acetate;dihydrate SMILES: [Li+].CC(=O)[O-].O.O
Lithium Acetate, Crystal, Spectrum™ Chemical
CAS: 6108-17-4 Molecular Formula: C2H7LiO4 Molecular Weight (g/mol): 102.01 InChI Key: IAQLJCYTGRMXMA-UHFFFAOYSA-M IUPAC Name: lithium(1+) acetate dihydrate SMILES: [Li+].O.O.CC([O-])=O
Lithium acetate dihydrate, Reagent Grade, Solstice
CAS: 6108-17-4 Molecular Formula: C2H7LiO4 Molecular Weight (g/mol): 102.01 MDL Number: MFCD00066949 InChI Key: IAQLJCYTGRMXMA-UHFFFAOYSA-M Synonym: Acetic acid lithium salt IUPAC Name: lithium(1+) acetate dihydrate SMILES: [Li+].O.O.CC([O-])=O