Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium acetate dihydrate, Reagent Grade, Solstice

Catalog No. 6003075 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
600-30-75 250 g
60-030-74 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 600-30-75 Supplier Solstice Supplier No. L6883250G

missing translation for 'validMainframeMsg'

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6108-17-4
Molecular Formula C2H7LiO4
Molecular Weight (g/mol) 102.01
MDL Number MFCD00066949
InChI Key IAQLJCYTGRMXMA-UHFFFAOYSA-M
Synonym Acetic acid lithium salt
IUPAC Name lithium(1+) acetate dihydrate
SMILES [Li+].O.O.CC([O-])=O

Specifications

CAS Max % ≤100.00
Physical Form Solid
Quantity 250 g
Beilstein 3564320
Grade Reagent
Packaging Plastic Bottle
Density 1.3 g/cm3
Precautionary Statement P281
RTECSNumber 208-914-3
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.