accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 611-01-8

IUPAC Name: 2,4-dimethylbenzoic acid
Synonyms: benzoic acid, 2,4-dimethyl-
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: BKYWPNROPGQIFZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C(O)=O)C(C)=C1
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (3)
  • (2)