Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylbenzoic acid, 98%

Catalog No. AC160320500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160320500 50 g Glass bottle
AC160320100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC160320500 Supplier Thermo Scientific Chemicals Supplier No. 160320500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 611-01-8 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-01-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00002480
InChI Key BKYWPNROPGQIFZ-UHFFFAOYSA-N
Synonym benzoic acid, 2,4-dimethyl, 4-carboxy-1,3-dimethylbenzene, 2,4-dimethyl benzoic acid, m-xylene-4-carboxylic acid, 2,4-dimethyl-benzoic acid, 2,4-dimethylbenzoicacid, m-xylylic acid, pubchem15441, 2,4 dimethylbenzoic acid, benzoic acid,4-dimethyl
PubChem CID 11897
ChEBI CHEBI:64811
IUPAC Name 2,4-dimethylbenzoic acid
SMILES CC1=CC=C(C(O)=O)C(C)=C1

Specifications

Melting Point 123°C to 127°C
Color White
CAS Min % 97.5
CAS Max % 100.0
Quantity 50 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 98%
Linear Formula (CH3)2C6H3CO2H
Beilstein 09, 531
Boiling Point 267°C (727.0 mmHg)
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 2, 4-Dimethylbenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-246-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.