accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 611-10-9

IUPAC Name: ethyl 2-oxocyclopentane-1-carboxylate
Synonyms: ethyl 2-oxocyclopentane-1-carboxylate
Molecular Weight (g/mol): 156.18
Molecular Formula: C8H12O3
InChi Key: JHZPNBKZPAWCJD-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCCC1=O
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (3)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)