Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyl 2-oxocyclopentanecarboxylate

Catalog No. 16809225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-809-225G 25 g
16-809-2100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-809-225G Supplier Sigma Aldrich Supplier No. 16809225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Phase-transfer benzylation reaction of ethyl 2-oxocyclopentanecarboxylate with benzyl bromide in microreactor has been investigated
  • Ethyl 2-oxocyclopentanecarboxylate participates in cobalt (ii) schiffαs base complex catalyzed oxidation of primary and secondary alcohols.ethyl 2-oxocyclopentanecarboxylate was used in stereoselective synthesis of (+/-)-cis,cis-spiro[4.4]nonane-1,6-diol
  • It was also used in the synthesis of tanikolide

Specifications

CAS 611-10-9
Density 1.054 g/mL (at 25°C (literature))
Boiling Point 102°C to 104°C (11 mmHg)
Molecular Formula C8H12O3
Refractive Index n20/D 1.452 (literature)
Linear Formula (O=)C5H7CO2C2H5
MDL Number MFCD00001412
Quantity 25 g
Synonym 2-(Ethoxycarbonyl)cyclopentanone; Ethyl cyclopentanone-2-carboxylate
Molecular Weight (g/mol) 156.18
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.