accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 611-21-2

IUPAC Name: N,2-dimethylaniline
Synonyms: N-methyl-O-toluidine
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: GUAWMXYQZKVRCW-UHFFFAOYSA-N
SMILES: CNC1=CC=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results