Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-o-toluidine 95.0+%, TCI America™
SDP

Catalog No. M044525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M044525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M044525ML Supplier TCI America Supplier No. M044525ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 611-21-2
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00025627
InChI Key GUAWMXYQZKVRCW-UHFFFAOYSA-N
Synonym n-methyl-o-toluidine, 2,n-dimethylaniline, benzenamine, n,2-dimethyl, o-toluidine, n-methyl, n,o-dimethylaniline, o,n-dimethylaniline, n-methyl-o-methylaniline, unii-e41nz1zy58, 2-methylamino toluene, n-methyltoluidine
PubChem CID 69137
IUPAC Name N,2-dimethylaniline
SMILES CC1=CC=CC=C1NC

Specifications

Color Yellow
Boiling Point 208°C
Quantity 25 mL
UN Number 2810
Formula Weight 121.18
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Methyl-o-toluidine
EINECSNumber (3)-0187
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.