accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 611-75-6

IUPAC Name: hydrogen 2-(1-amino-1-cyclohexylethyl)-4,6-dibromoaniline chloride
Synonyms: hydrogen 2-(1-amino-1-cyclohexylethyl)-4,6-dibromoaniline chloride
Molecular Weight (g/mol): 412.59
Molecular Formula: C14H21Br2ClN2
InChi Key: SXIVGYTWSVJOLM-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(N)(C1CCCCC1)C1=CC(Br)=CC(Br)=C1N
Percent Purity 
  • (2)
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)