accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOOH

CAS RN 61166-00-5

IUPAC Name: 3-[2-(3-hydroxyphenoxy)ethoxy]phenol
Synonyms: 3-[2-(3-hydroxyphenoxy)ethoxy]phenol
Molecular Weight (g/mol): 246.26
Molecular Formula: C14H14O4
InChi Key: DBFHCPVZZSVFJL-UHFFFAOYSA-N
SMILES: OC1=CC(OCCOC2=CC(O)=CC=C2)=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results