Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3'-Ethylenedioxydiphenol, TCI America™
SDP

Catalog No. E044825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E044825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E044825G Supplier TCI America Supplier No. E044825G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61166-00-5
Molecular Formula C14H14O4
Molecular Weight (g/mol) 246.26
MDL Number MFCD00002268
InChI Key DBFHCPVZZSVFJL-UHFFFAOYSA-N
Synonym 1,2-Bis(3-hydroxyphenoxy)ethane, Ethylene Glycol Bis(3-hydroxyphenyl) Ether
PubChem CID 109057
IUPAC Name 3-[2-(3-hydroxyphenoxy)ethoxy]phenol
SMILES OC1=CC(OCCOC2=CC(O)=CC=C2)=CC=C1

Specifications

Melting Point 161°C
Quantity 25 g
Formula Weight 246.26
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Ethylenedioxydiphenol
EINECSNumber (3)-2761
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.