accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61189-99-9

IUPAC Name: 2,2-diethoxyacetamide
Synonyms: 2,2-diethoxyacetamide
Molecular Weight (g/mol): 147.17
Molecular Formula: C6H13NO3
InChi Key: DSKGWFQMJHBHRT-UHFFFAOYSA-N
SMILES: CCOC(OCC)C(N)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results