Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Diethoxyacetamide 99%, Thermo Scientific™

Catalog No. AC279880500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC279880500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC279880500 Supplier Thermo Scientific Chemicals Supplier No. 279880500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61189-99-9
Molecular Formula C6H13NO3
Molecular Weight (g/mol) 147.17
MDL Number MFCD00044023
InChI Key DSKGWFQMJHBHRT-UHFFFAOYSA-N
Synonym acetamide,2,2-diethoxy, 2,2-diethoxy-acetamide, diethoxyacetamide, acmc-20a5q0, acetamide, 2,2-diethoxy, glyoxylic amide diethyl acetal, 2,2-diethoxyacetamide
PubChem CID 100765
IUPAC Name 2,2-diethoxyacetamide
SMILES CCOC(OCC)C(N)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 77°C to 80°C
Color White
Infrared Spectrum Authentic
Linear Formula (C2H5O)2CHC(O)NH2
Quantity 50 g
Formula Weight 147.17
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 2, 2-Diethoxyacetamide, 99%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.