accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 612-13-5

IUPAC Name: 2-(chloromethyl)benzonitrile
Synonyms: 2-cyanobenzyl chloride
Molecular Weight (g/mol): 151.59
Molecular Formula: C8H6ClN
InChi Key: ZSHNOXOGXHXLAV-UHFFFAOYSA-N
SMILES: ClCC1=CC=CC=C1C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results