accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 612-22-6

H3CNOO

CAS RN 612-22-6

IUPAC Name: 1-ethyl-2-nitrobenzene
Synonyms: 2-ethylnitrobenzene
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: PXWYZLWEKCMTEZ-UHFFFAOYSA-N
SMILES: CCC1=CC=CC=C1[N+]([O-])=O
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)