accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 612-69-1

IUPAC Name: 3,3',5,5'-tetramethoxy-[1,1'-biphenyl]-4,4'-diol
Synonyms: 3,3',5,5'-tetramethoxy-[1,1'-biphenyl]-4,4'-diol
Molecular Weight (g/mol): 306.31
Molecular Formula: C16H18O6
InChi Key: NRTOUGPMXLJETQ-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1O)C1=CC(OC)=C(O)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results