Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrocoerulignone, MP Biomedicals

Catalog No. ICN211955 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN211955 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN211955 Supplier MP Biomedicals, Inc Supplier No. 0521195501

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 612-69-1
Molecular Formula C16H18O6
Molecular Weight (g/mol) 306.314
InChI Key NRTOUGPMXLJETQ-UHFFFAOYSA-N
Synonym 3,3',5,5'-tetramethoxy 1,1'-biphenyl-4,4'-diol, hydrocoerulignone, 4-4-hydroxy-3,5-dimethoxyphenyl-2,6-dimethoxyphenol, 1,1'-biphenyl, 3,3',5,5'-tetramethoxy-4,4'-dihydroxy, hydrocerulignone, ghl.pd_mitscher_leg0.943, 3,3',5,5'-tetramethoxy biphenyl-4,4'-diol, 3,3',5,5'-tetramethoxy-1,1'-biphenyl-4,4'-diol, 4-4-hydroxy-3,5-dimethoxy-phenyl-2,6-dimethoxy-phenol
PubChem CID 256604
IUPAC Name 4-(4-hydroxy-3,5-dimethoxyphenyl)-2,6-dimethoxyphenol
SMILES COC1=CC(=CC(=C1O)OC)C2=CC(=C(C(=C2)OC)O)OC

Specifications

Quantity 1 g
Chemical Name or Material Hydrocoerulignone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.