accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 612-96-4

IUPAC Name: 2-phenylquinoline
Synonyms: 2-phenylquinoline
Molecular Weight (g/mol): 205.26
Molecular Formula: C15H11N
InChi Key: FSEXLNMNADBYJU-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC=C2C=CC=CC2=N1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (1)
Quantity 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (5)
  • (3)
  • (1)