Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylquinoline, 99+%

Catalog No. AAH3192006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3192006 5 g
AAH3192003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3192006 Supplier Thermo Scientific Chemicals Supplier No. H3192006
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 612-96-4 | C15H11N | 205.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-96-4
Molecular Formula C15H11N
Molecular Weight (g/mol) 205.26
MDL Number MFCD00011568
InChI Key FSEXLNMNADBYJU-UHFFFAOYSA-N
Synonym quinoline, 2-phenyl, 2-phenyl quinoline, .alpha.-phenylquinoline, 2-phenyl-quinoline, quinoline, phenyl, chembl89786, a-phenylquinoline, quinoline,phenyl, phenylquinoline, pubchem15940
PubChem CID 71545
IUPAC Name 2-phenylquinoline
SMILES C1=CC=C(C=C1)C1=CC=C2C=CC=CC2=N1

Specifications

Melting Point 82°C to 84°C
Boiling Point 363°C
Quantity 5 g
Formula Weight 205.26
Percent Purity ≥99%
Chemical Name or Material 2-Phenylquinoline
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-326-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.